  • Search Results
  • Antibacterial active compounds from Hypericum ascyron L. induce bacterial cell death through apoptosis pathway.
Antibacterial active compounds from Hypericum ascyron L. induce bacterial cell death through apoptosis pathway.
European journal of medicinal chemistry (2015-04-29)
Xiu-Mei Li, Xue-Gang Luo, Chuan-Ling Si, Nan Wang, Hao Zhou, Jun-Fang He, Tong-Cun Zhang
ABSTRACT
Hypericum ascyron L. has been used as a traditional medicine for the treatment of wounds, swelling, headache, nausea and abscesses in China for thousands of years. However, modern pharmacological studies are still necessary to provide a scientific basis to substantiate their traditional use. In this study, the mechanism underlying the antimicrobial effect of the antibacterial activity compounds from H. ascyron L. was investigated. Bioguided fractionation of the extract from H. ascyron L. afforded antibacterial activity fraction 8. The results of cup plate analysis and MTT assay showed that the MIC and MBC of fraction 8 is 5 mg/mL. Furthermore, using Annexin V-FITC/PI, TUNEL labeling and DNA gel electrophoresis, we found that cell death with apoptosis features similar to those in eucaryon could be induced in bacteria strains after exposure to the antibacterial activity compounds from H. ascyron L. at moderate concentration. In addition, we further found fraction 8 could disrupt the cell membrane potential indicate that fraction 8 exerts pro-apoptotic effects through a membrane-mediated apoptosis pathway. Finally, quercetin and kaempferol 3-O-β-(2″-acetyl)-galactopyranoside, were identified from fraction 8 by means of Mass spectrometry and Nuclear magnetic resonance. To our best knowledge, this study is the first to show that Kaempferol 3-O-β-(2″-acetyl)-galactopyranoside coupled with quercetin had significant antibacterial activity via apoptosis pathway, and it is also the first report that Kaempferol 3-O-β-(2″-acetyl)-galactopyranoside was found in clusiacea. Our data might provide a rational base for the use of H. ascyron L. in clinical, and throw light on the development of novel antibacterial drugs.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D