  • Search Results
  • Calcium phosphate nanoparticles carrying BMP-7 plasmid DNA induce an osteogenic response in MC3T3-E1 pre-osteoblasts.
Calcium phosphate nanoparticles carrying BMP-7 plasmid DNA induce an osteogenic response in MC3T3-E1 pre-osteoblasts.
Journal of biomedical materials research. Part A (2015-06-23)
Chrystalleni Hadjicharalambous, Diana Kozlova, Viktoriya Sokolova, Matthias Epple, Maria Chatzinikolaidou
ABSTRACT
Functionalized calcium phosphate nanoparticles with osteogenic activity were prepared. Polyethyleneimine-stabilized calcium phosphate nanoparticles were coated with a shell of silica and covalently functionalized by silanization with thiol groups. Between the calcium phosphate surface and the outer silica shell, plasmid DNA which encoded either for bone morphogenetic protein 7 (BMP-7) or for enhanced green fluorescent protein was incorporated as cargo. The plasmid DNA-loaded calcium phosphate nanoparticles were used for the transfection of the pre-osteoblastic MC3T3-E1 cells. The cationic nanoparticles showed high transfection efficiency together with a low cytotoxicity. Their potential to induce an osteogenic response by transfection was demonstrated by measuring the alkaline phosphatase (ALP) activity and calcium deposition with alizarin red staining. The expression of the osteogenic markers Alp, Runx2, ColIa1 and Bsp was investigated by means of real-time quantitative polymerase chain reaction. It was shown that phBMP-7-loaded nanoparticles can provide a means of transient transfection and localized production of BMP-7 in MC3T3-E1 cells, with a subsequent increase of two osteogenic markers, specifically ALP activity and calcium accumulation in the extracellular matrix. Future strategies to stimulate bone regeneration focus into enhancing transfection efficiency and achieving higher levels of BMP-7 produced by the transfected cells.
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium phosphate, purum p.a., ≥96.0% (calc. as Ca3(PO4)2, KT)
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Trimethoxysilane, 95%
Sigma-Aldrich
Sodium 3-mercapto-1-propanesulfonate, technical grade, 90%
Sigma-Aldrich
1,2,4,5-Tetramethylbenzene, 98%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N