  • Search Results
  • Design Guidelines for High-Performance Particle-Based Photoanodes for Water Splitting: Lanthanum Titanium Oxynitride as a Model.
Design Guidelines for High-Performance Particle-Based Photoanodes for Water Splitting: Lanthanum Titanium Oxynitride as a Model.
ChemSusChem (2015-09-12)
Steve Landsmann, Alexandra E Maegli, Matthias Trottmann, Corsin Battaglia, Anke Weidenkaff, Simone Pokrant
ABSTRACT
Semiconductor powders are perfectly suited for the scalable fabrication of particle-based photoelectrodes, which can be used to split water using the sun as a renewable energy source. This systematic study is focused on variation of the electrode design using LaTiO2 N as a model system. We present the influence of particle morphology on charge separation and transport properties combined with post-treatment procedures, such as necking and size-dependent co-catalyst loading. Five rules are proposed to guide the design of high-performance particle-based photoanodes by adding or varying several process steps. We also specify how much efficiency improvement can be achieved using each of the steps. For example, implementation of a connectivity network and surface area enhancement leads to thirty times improvement in efficiency and co-catalyst loading achieves an improvement in efficiency by a factor of seven. Some of these guidelines can be adapted to non-particle-based photoelectrodes.
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%