  • Search Results
  • Application of GC-MS coupled with chemometrics for scanning serum metabolic biomarkers from renal fibrosis rat.
Application of GC-MS coupled with chemometrics for scanning serum metabolic biomarkers from renal fibrosis rat.
Biochemical and biophysical research communications (2015-04-18)
Shao Liu, Ji-Shi Liu, Ren-Na Luo, Hui Xu, Wei-Ru Zhang, Jie Meng, Yi-Zeng Liang, Li-Jian Tao
ABSTRACT
Renal interstitial fibrosis closely relates to chronic kidney disease and is regarded as the final common pathway in most cases of end-stage renal disease. Metabolomic biomarkers can facilitate early diagnosis and allow better understanding of the pathogenesis underlying renal fibrosis. Gas chromatography-mass spectrometry (GC/MS) is one of the most promising techniques for identification of metabolites. However, the existence of the background, baseline offset, and overlapping peaks makes accurate identification of the metabolites unachievable. In this study, GC/MS coupled with chemometric methods was successfully developed to accurately identify and seek metabolic biomarkers for rats with renal fibrosis. By using these methods, seventy-six metabolites from rat serum were accurately identified and five metabolites (i.e., urea, ornithine, citric acid, galactose, and cholesterol) may be useful as potential biomarkers for renal fibrosis.
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Sigma-Aldrich
Heptadecanoic acid, ≥98%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur