  • Search Results
  • Fatty acids attached to all-trans-astaxanthin alter its cis-trans equilibrium, and consequently its stability, upon light-accelerated autoxidation.
Fatty acids attached to all-trans-astaxanthin alter its cis-trans equilibrium, and consequently its stability, upon light-accelerated autoxidation.
Food chemistry (2015-10-17)
Wouter J C de Bruijn, Yannick Weesepoel, Jean-Paul Vincken, Harry Gruppen
ABSTRACT
Fatty acid esterification, common in naturally occurring astaxanthin, has been suggested to influence both colour stability and degradation of all-trans-astaxanthin. Therefore, astaxanthin stability was studied as influenced by monoesterification and diesterification with palmitate. Increased esterification decelerated degradation of all-trans-astaxanthin (RP-UHPLC-PDA), whereas, it had no influence on colour loss over time (spectrophotometry). This difference might be explained by the observation that palmitate esterification influenced the cis-trans equilibrium. Free astaxanthin produced larger amounts of 9-cis isomer whereas monopalmitate esterification resulted in increased 13-cis isomerization. The molar ratios of 9-cis:13-cis after 60min were 1:1.7 (free), 1:4.8 (monopalmitate) and 1:2.6 (dipalmitate). The formation of 9-cis astaxanthin, with its higher molar extinction coefficient than that of all-trans-astaxanthin, might compensate for colour loss induced by conjugated double bond cleavage. As such, it was concluded that spectrophotometry is not an accurate measure of the degradation of the all-trans-astaxanthin molecule.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Astaxanthin, ≥97% (HPLC), from Blakeslea trispora
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Hydriodic acid, puriss. p.a., ≥67% (T)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)