  • Search Results
  • Optimization of supercritical fluid extraction of essential oils and fatty acids from flixweed (Descurainia Sophia L.) seed using response surface methodology and central composite design.
Optimization of supercritical fluid extraction of essential oils and fatty acids from flixweed (Descurainia Sophia L.) seed using response surface methodology and central composite design.
Journal of food science and technology (2015-07-04)
Katayoun Mahdavi Ara, Mina Jowkarderis, Farhad Raofie
ABSTRACT
Essential oils and fatty acids of Descurainia sophia L. seed were obtained by supercritical CO2 extraction and steam distillation methods. The effect of different parameters such as pressure, temperature, modifier volume, dynamic and static extraction timeon the extraction yield were optimized using a central composite design after a 2 (n-1) fractional factorial design. The results showed that under the pressure of 355 bar, temperature of 65 °C, methanol volume of 150 μL, dynamic and static extraction times of 35 and 10 min, respectively, the major components were methyl linoleate (18.2 %), camphor (12.32 %), cis-thujone (11.3 %) and trans-caryophyllene (9.17 %). The results indicated that by using the proper conditions, the supercritical fluid extraction is more selective than the steam distillation method. Extraction yields based on supercritical fluid extraction varied in the range of 0.68 to 17.1 % (w/w), and the extraction yield based on the steam distillation was 0.25 % (v/w).
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), Customized plastic drum
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), One-time steel-plastic (SP) drum
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Methanol, NMR reference standard
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid