  • Search Results
  • Gamma-irradiation of deglycerolized red cells does not significantly affect in vitro quality.
Gamma-irradiation of deglycerolized red cells does not significantly affect in vitro quality.
Vox sanguinis (2015-05-09)
K M Winter, L Johnson, R G Webb, D C Marks
ABSTRACT
Red cells frozen with glycerol may require gamma-irradiation after thawing and deglycerolization for transfusion to at-risk patients. Both freezing and irradiation are known to cause red cell damage. However, the effect of irradiation on the quality of deglycerolized red cells and the optimal shelf life of such a component is currently unknown. Red cells (<7 days) were pooled, split and glycerolized using an ACP-215 automated cell washer (n = 12 pairs) and frozen at -80°C. Red cells were thawed, deglycerolized and resuspended in SAG-M. One of each pair was gamma-irradiated, while the other served as a control. Products were stored at 2-6°C and sampled for in vitro testing immediately after irradiation, and at 24 and 48 h postirradiation. Irradiation of deglycerolized red cells led to a >1·5-fold increase in extracellular potassium, compared to control units at 24 and 48 h postirradiation. Other parameters, including haemolysis, were not significantly affected by irradiation postdeglycerolization. Deglycerolized, irradiated red cells had increased supernatant potassium, but remained of acceptable quality for 24 h postirradiation.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis