  • Search Results
  • A Gelatinases-targeting scFv-based Fusion Protein Shows Enhanced Antitumour Activity with Endostar against Hepatoma.
A Gelatinases-targeting scFv-based Fusion Protein Shows Enhanced Antitumour Activity with Endostar against Hepatoma.
Basic & clinical pharmacology & toxicology (2015-01-24)
Ruijuan Gao, Liang Li, Boyang Shang, Chunyan Zhao, Weijin Sheng, Diandong Li
ABSTRACT
Gelatinases play important roles in tumour invasion and metastasis and are thus considered promising targets for cancer therapy. In this study, a new single-chain variable fragment (scFv)-based fusion protein Fv-LDP, composed of the anti-gelatinases scFv and lidamycin apoprotein (LDP), was prepared, and its combination with angiogenesis inhibitor Endostar was then investigated. The fusion protein Fv-LDP specifically bound to various tumour cells, and its binding capability to human pulmonary giant cell carcinoma (PG) cells was higher than that of LDP. Fv-LDP inhibited the expression and secretion of gelatinases and could be internalized into tumour cells via endocytosis. Fv-LDP also suppressed the growth of human hepatoma cells and murine hepatoma 22 transplanted in Kunming mice in various degrees. In addition, Endostar could enhance the synergistic or additive inhibition of Fv-LDP on the growth, migration or invasion of human hepatoma cells shown by a colony formation assay and a transwell-based migration or invasion assay, respectively. In vivo, Fv-LDP/Endostar combination showed a significantly synergistic effect on the growth of a human hepatoma xenograft, with an inhibition rate of 80.8% compared with the Fv-LDP (44.1%) or Endostar (8.9%)-treated group. The above-mentioned results indicate that the fusion protein Fv-LDP is effective against transplantable hepatoma in mice and human hepatoma xenografts in athymic mice. Moreover, Endostar can potentiate the inhibition effect of Fv-LDP on the growth of human hepatoma cells and xenografts. These data will provide a new combined strategy for improving the therapeutic efficacy of treatments for hepatoma or other gelatinase-overexpressing tumours.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
1,3-Propanediol, 98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture