  • Search Results
  • Modulation of biomechanical properties of hyaluronic acid hydrogels by crosslinking agents.
Modulation of biomechanical properties of hyaluronic acid hydrogels by crosslinking agents.
Journal of biomedical materials research. Part A (2015-02-19)
Sung Chul Choi, Mi Ae Yoo, Su Yeon Lee, Hyun Ji Lee, Dong Hoon Son, Jessica Jung, Insup Noh, Chan-Wha Kim
ABSTRACT
Modulation of both mechanical properties and biocompatibilities of hyaluronic acid (HA) hydrogels is very importance for their applications in biomaterials. Pure HA solution was converted into a hydrogel by using butanediol diglycidyl ether (BDDE) as a crosslinking agent. Mechanical properties of the HA hydrogels have been evaluated by adding up different amount of BDDEs. While the mechanical properties of the obtained HA hydrogels were evaluated by measuring their crosslinking degrees, elastic modulus and viscosity, their in vitro biocompatibilities were done by measuring the degrees of anti-inflammatory reactions, cell viabilities and cytotoxicity. The degrees of anti-inflammatory reactions were determined by measuring the amount of nitric oxides (NOs) released from lipopolysaccharide(LPS)(+)-induced macrophages; cell viability was evaluated by observing differences in the behaviors of fibroblasts covered with the HA hydrogels, compared with those covered with the films of Teflon and Latex. Cytotoxicity of the HA hydrogels was also evaluated by measuring the degrees of viability of the cells exposed on the extracts of the HA hydrogels over those of Teflon, Latex and pure HA solutions by the assays of thiazoly blue tetrazolium bromide (MTT), neutral reds, and bromodeoxyuridine (BrdU). The results showed that employment of BDDEs beyond critical amounts showed lower biocompatibility of the crosslinked HA hydrogels but higher crosslinking degrees and mechanical properties, indicating the importance of controlling the HA concentrations, BDDE amounts and their reaction times for the synthesis of the crosslinked HA hydrogels for their clinical applications as biomaterials.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)