  • Search Results
  • Multi-beam laser-induced hydrodynamic shock waves used for delivery of microparticles and liquids in skin.
Multi-beam laser-induced hydrodynamic shock waves used for delivery of microparticles and liquids in skin.
Lasers in surgery and medicine (2015-09-27)
Andrey V Belikov, Alexei V Skrypnik, Ksenia V Shatilova, Valery V Tuchin
ABSTRACT
Laser radiation is often used to provide micro and nanoparticle delivery into the skin for medical and cosmetic purposes. This technique inherently has limited speed and effective penetration. We proposed and investigated a new method of rapid delivery of solid microparticles, nanoparticles and liquids into tissue through multiple microchannels created by a fractional laser microablation (FLMA) using Er:YAG-laser. The dependence of microchannel depth on laser pulse temporal structure and number of pulses and dermal coloration changes are studied in this paper. Microchannels created in the porcine skin in vitro by a fractional Er:YAG-laser were used to deliver Zirconium oxide (ZrO2) microparticles or hydrocortisone solution. Each laser pulse consisted of subpulses. Number of laser pulses (Np) and subpulses (Nsp) can be adjusted. The enhancement of delivery is expected due to hydrodynamic impact of laser pulse on the layer of the aqueous suspension of the particles or hydrocortisone solution placed on the skin surface. For color investigation, we used standard CIE Lab parameter analysis. The relationship between microchannel depth in the skin and number of laser pulses and subpulses was established. We found that free filling of microchannels with ZrO2-particle suspension has a low speed of ∼4 × 10(-5)  mm/s. Particle delivery into microchannels induced by the hydrodynamic shock waves generated by Er:YAG-laser pulses is carried out with a high speed of 28.5 mm/s. We also found that skin color at ZrO2 -particle delivery differs from color of the intact skin, namely: the parameter L, which characterizes the "lightness" increased by 9 ± 1%; parameter a, which characterizes the "redness" decreased by 38 ± 4%; and parameter b, which characterizes the "yellowness" decreased by 21 ± 2%. The effective delivery of hydrocortisone was demonstrated using fluorescence method technique. Multi-beam laser-induced hydrodynamic shock waves generated by Er:YAG-laser pulses on the layer of the aqueous suspension of the particles or solution of a high molecular weight drug placed on the skin can be used for their rapid delivery into the skin.
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Yttrium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Yttrium, wire reel, 0.05m, diameter 1.0mm, square section, 99.9%
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Sigma-Aldrich
Potassium chloride, Vetec, reagent grade, 99%
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%