  • Search Results
  • Experimental Verification of the Effects on Normal Domestic Cats by Feeding Prescription Diet for Decreasing Stress.
Experimental Verification of the Effects on Normal Domestic Cats by Feeding Prescription Diet for Decreasing Stress.
Journal of applied animal welfare science : JAAWS (2015-02-14)
Kazuki Miyaji, Maki Kato, Nobuyo Ohtani, Mitsuaki Ohta
ABSTRACT
The objective of this study was to evaluate the effects of diet on the feline stress response by measuring plasma and urinary cortisol. A study diet was developed with a unique combination of nutrients that supports the management of stressful situations. The specific formulation of the diet included alpha-casozepine, which is believed to have an anxiolytic effect, and tryptophan supplementation. Tryptophan is the precursor for the synthesis of the neurotransmitter serotonin. Twenty-one indoor cats were fed with the study diet (n = 10) or a control diet (n = 11) for 8 weeks, after which physiological responses were evaluated. The study diet significantly increased the ratio of plasma tryptophan to large neutral amino acids and decreased urinary cortisol concentrations after being consumed daily for 8 weeks, but there was no effect on plasma cortisol levels following a stressful event (veterinary examination and blood draw). Further studies, such as behavioral analyses, are needed to clarify the effects of the study diet.
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
SAFC
Taurine
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, Vetec, reagent grade, 99%
Sigma-Aldrich
Choline chloride, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Taurine, Vetec, reagent grade, ≥99%
Sigma-Aldrich
Calcium sulfate, Vetec, reagent grade
Sigma-Aldrich
DL-Methionine, Vetec, reagent grade, 98%