  • Search Results
  • Dual dispersive liquid-liquid microextraction for determination of phenylpropenes in oils by gas chromatography-mass spectrometry.
Dual dispersive liquid-liquid microextraction for determination of phenylpropenes in oils by gas chromatography-mass spectrometry.
Journal of chromatography. A (2015-08-09)
Chia-Ju Tsai, Jih-Heng Li, Chia-Hsien Feng
ABSTRACT
A novel, simple and quick sample preparation method was developed and used for pre-concentration and extraction of six phenylpropenes, including anethole, estragole, eugenol, methyl eugenol, safrole and myristicin, from oil samples by dual dispersive liquid-liquid microextraction. Gas chromatography-mass spectrometry was used for determination and separation of compounds. Several experimental parameters affecting extraction efficiency were evaluated and optimized, including forward-extractant type and volume, surfactant type and concentration, water volume, and back-extractant type and volume. For all analytes (10-1000ng/mL), the limits of detection (S/N≧3) ranged from 1.0 to 3.0ng/mL; the limits of quantification (S/N≧10) ranged from 2.5 to 10.0ng/mL; and enrichment factors ranged from 3.2 to 37.1 times. Within-run and between-run relative standard deviations (n=6) were less than 2.61% and less than 4.33%, respectively. Linearity was excellent with determination coefficients (r(2)) above 0.9977. The experiments showed that the proposed method is a simple, effective, and environmentally friendly method of analyzing phenylpropenes in oil samples.
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Vetec, reagent grade, 96%
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), One-time steel-plastic (SP) drum
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
SAFC
Hexadecyltrimethylammonium bromide
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), Customized plastic drum
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%