Responsive foams for nanoparticle delivery.
Colloids and surfaces. B, Biointerfaces (2015-06-21)
Christina Tang, Edward Xiao, Patrick J Sinko, Zoltan Szekely, Robert K Prud'homme
ABSTRACT
We have developed responsive foam systems for nanoparticle delivery. The foams are easy to make, stable at room temperature, and can be engineered to break in response to temperature or moisture. Temperature-responsive foams are based on the phase transition of long chain alcohols and could be produced using medical grade nitrous oxide as a propellant. These temperature-sensitive foams could be used for polyacrylic acid (PAA)-based nanoparticle delivery. We also discuss moisture-responsive foams made with soap pump dispensers. Polyethylene glycol (PEG)-based nanoparticles or PMMA latex nanoparticles were loaded into Tween 20 foams and the particle size was not affected by the foam formulation or foam break. Using biocompatible detergents, we anticipate this will be a versatile and simple approach to producing foams for nanoparticle delivery with many potential pharmaceutical and personal care applications.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Propylene glycol, FCC, FG
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octadecanol, ReagentPlus®, 99%
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1-Octadecanol, 95%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Nitrous oxide, 99%
Sigma-Aldrich
1-Hexadecanol, ≥99%
Sigma-Aldrich
1-Hexadecanol, 95%
Sigma-Aldrich
Potassium hydroxide on alumina
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free