  • Search Results
  • Porous gelatin/tricalcium phosphate/genipin composites containing lumbrokinase for bone repair.
Porous gelatin/tricalcium phosphate/genipin composites containing lumbrokinase for bone repair.
Bone (2015-05-03)
Yuan-Tsung Fu, Shi-Yuan Sheu, Yueh-Sheng Chen, Kuo-Yu Chen, Chun-Hsu Yao
ABSTRACT
Bone cell activities are very important in bone remodeling. This study investigates the effects of lumbrokinase on bone cell activities in cultures. Moreover, a biodegradable composite (GGT) containing genipin-crosslinked gelatin and β-tricalcium phosphate was prepared to carry lumbrokinase (GGTLK). Rat calvarial bone defects were filled with GGT and GGTLK composites. Bone healing was monitored in vivo by bioluminescence imaging and micro-CT. Lumbrokinase was found to have a dose-dependent effect on bone cell activities. Low concentrations (<1μg/ml) of lumbrokinase increased the viability, total alkaline phosphatase activity and mobility of osteoblasts, the number of total calcified nodules and the expression of osteopontin and osteocalcin; however, they considerably reduced the total tartrate-resistant acid phosphatase activity of osteoclasts. IVIS images revealed a stronger fluorescent signal in GGTLK-treated animals than in GGT-treated animals. Micro-CT analysis revealed that GGTLK induced more new bone formation than did GGT. These observations suggest that lumbrokinase released from GGTLK composite can enhance bone tissue regeneration.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Aluminum sulfate, 99.99% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
L-Ascorbic acid, Vetec, reagent grade, 99%
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)