  • Search Results
  • Chemiluminescence immunoassay using magnetic nanoparticles with targeted inhibition for the determination of ochratoxin A.
Chemiluminescence immunoassay using magnetic nanoparticles with targeted inhibition for the determination of ochratoxin A.
Talanta (2015-06-07)
Sumin Kim, H B Lim
ABSTRACT
In this work, a chemiluminescence (CL) immunoassay with targeted inhibition was developed for the determination of toxins in food products. For sample treatment, amine-functionalized magnetic nanoparticles (MNPs) were synthesized to extract target molecules, and horseradish peroxidase (HRP) tagged on an antibody was used as a label for CL reaction. In particular, amine-targeted inhibition using aldehyde, i.e., specifically capping the amine with an alkyl group, was developed for a non-specific extraction platform to lower background and improve signal-to-background ratio. For demonstration, ochratoxin A (OTA) was determined in rice using a lab-built drop-type chemiluminescence (DCL) system with luminol-H2O2 reagent. The obtained limit of detection was 1.39 pg mL(-1), which was about 7.3 times better than that of ELISA. Recovery of the method in the range of 87-99% was observed, which was compared with ELISA.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
4-Iodophenol, 99%
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N