  • Search Results
  • Effects of 24 CYP2D6 Variants Found in the Chinese Population on the Metabolism of Risperidone.
Effects of 24 CYP2D6 Variants Found in the Chinese Population on the Metabolism of Risperidone.
Pharmacology (2015-11-07)
Zhen-He Wang, Yun-Yun Zhan, Yun-Xuan Li, Cheng-Cheng Yang, Jie Cai, Da-Peng Dai, Guo-Xin Hu, Jian-Ping Cai
ABSTRACT
Cytochrome P450 (CYP450) 2D6 is an important member of the P450 enzyme superfamily and responsible for clearing 25% of clinically important drugs. The aim of this study was to assess the catalytic characteristics of 24 CYP2D6 allelic isoforms found in the Chinese population and their effects on the metabolism of risperidone in vitro. Insect microsomes expressing wild-type CYP2D6 and 24 CYP2D6 allelic variants were incubated with 20-1,000 μmol/l risperidone for 40 min at 37°C. After termination, risperidone and 9-OH risperidone, the metabolite of risperidone, were precipitated and used for signal collection by ultra-performance liquid-chromatography tandem mass spectrometry. Among 24 CYP2D6 variants tested, 2 variants (CYP2D6*92 and CYP2D6*96) were found to be with no detectable activity. Two variants (E215K and R440C) exhibited higher intrinsic clearance values than the wild-type protein, while the remaining 20 CYP2D6 allelic variants exhibited significantly decreased clearance values (2.01-87.56%) compared to CYP2D6*1. These findings suggest that more attention should be directed to subjects carrying these infrequent CYP2D6 alleles when administering risperidone in the clinic. This is the first report of all these novel alleles for risperidone metabolism, providing fundamental data for further clinical studies on CYP2D6 alleles.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, configured for PerkinElmer 8900, configured for Polygen, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile