  • Search Results
  • Synthesis and biological evaluation of nigranoic acid esters as novel human neutrophil elastase inhibitors.
Synthesis and biological evaluation of nigranoic acid esters as novel human neutrophil elastase inhibitors.
Natural product research (2015-01-07)
Guoli Huang, Li Feng, Bo Liu, Yi He, Yiming Li, Yegao Chen
ABSTRACT
Human neutrophil elastase (HNE) has been implicated as a major contributor in the pathogenesis of diseases, such as lung disorders and other inflammatory diseases. A series of 12 new nigranoic acid esters were regioselectively synthesised in good yields and evaluated for HNE inhibitory activity. Nigranoic acid exhibited significant inhibitory activity against HNE with the IC50 value of 3.77 μM, and six esters displayed considerable inhibitory effects on HNE with IC50 values in the range of 2.61-8.95 μM. The nigranoic acid esters having phenyls substituted with bromine and trimethoxyls (3h and 3b) showed stronger inhibitory activity on HNE than nigranoic acid.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
L-Proline p-nitroanilide trifluoroacetate salt, prolyl aminopeptidase substrate
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)