  • Search Results
  • Vitamin C prevents the endothelial dysfunction induced by acute ethanol intake.
Vitamin C prevents the endothelial dysfunction induced by acute ethanol intake.
Life sciences (2015-09-20)
Ulisses V Hipólito, Glaucia E Callera, Janaina A Simplicio, Bruno S De Martinis, Rhian M Touyz, Carlos R Tirapelli
ABSTRACT
Investigate the effect of ascorbic acid (vitamin C) on the endothelial dysfunction induced by acute ethanol intake. Ethanol (1g/kg; p.o. gavage) effects were assessed within 30min in male Wistar rats. Ethanol intake decreased the endothelium-dependent relaxation induced by acetylcholine in the rat aorta and treatment with vitamin C (250mg/kg; p.o. gavage, 5days) prevented this response. Ethanol increased superoxide anion (O2(-)) generation and decreased aortic nitrate/nitrite levels and these responses were not prevented by vitamin C. Superoxide dismutase (SOD) and catalase (CAT) activities as well as hydrogen peroxide (H2O2) and reduced glutathione (GSH) levels were not affected by ethanol. RhoA translocation as well as the phosphorylation levels of protein kinase B (Akt), eNOS (Ser(1177) or Thr(495) residues), p38MAPK, SAPK/JNK and ERK1/2 was not affected by ethanol intake. Vitamin C increased SOD activity and phosphorylation of Akt, eNOS (Ser(1177) residue) and p38MAPK in aortas from both control and ethanol-treated rats. Incubation of aortas with tempol prevented ethanol-induced decrease in the relaxation induced by acetylcholine. Ethanol (50mM/1min) increased O2(-) generation in cultured aortic vascular smooth muscle cells (VSMC) and vitamin C did not prevent this response. In endothelial cells, vitamin C prevented the increase on ROS generation and the decrease in the cytosolic NO content induced by ethanol. Our study provides novel evidence that vitamin C prevents the endothelial dysfunction induced by acute ethanol intake by a mechanism that involves reduced ROS generation and increased NO availability in endothelial cells.
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, Vetec, reagent grade, 99%