  • Search Results
  • Loss of a Conserved tRNA Anticodon Modification Perturbs Plant Immunity.
Loss of a Conserved tRNA Anticodon Modification Perturbs Plant Immunity.
PLoS genetics (2015-10-23)
Vicente Ramírez, Beatriz Gonzalez, Ana López, María José Castelló, María José Gil, Graham J Etherington, Bo Zheng, Peng Chen, Pablo Vera
ABSTRACT
tRNA is the most highly modified class of RNA species, and modifications are found in tRNAs from all organisms that have been examined. Despite their vastly different chemical structures and their presence in different tRNAs, occurring in different locations in tRNA, the biosynthetic pathways of the majority of tRNA modifications include a methylation step(s). Recent discoveries have revealed unprecedented complexity in the modification patterns of tRNA, their regulation and function, suggesting that each modified nucleoside in tRNA may have its own specific function. However, in plants, our knowledge on the role of individual tRNA modifications and how they are regulated is very limited. In a genetic screen designed to identify factors regulating disease resistance and activation of defenses in Arabidopsis, we identified SUPPRESSOR OF CSB3 9 (SCS9). Our results reveal SCS9 encodes a tRNA methyltransferase that mediates the 2´-O-ribose methylation of selected tRNA species in the anticodon loop. These SCS9-mediated tRNA modifications enhance during the course of infection with the bacterial pathogen Pseudomonas syringae DC3000, and lack of such tRNA modification, as observed in scs9 mutants, severely compromise plant immunity against the same pathogen without affecting the salicylic acid (SA) signaling pathway which regulates plant immune responses. Our results support a model that gives importance to the control of certain tRNA modifications for mounting an effective immune response in Arabidopsis, and therefore expands the repertoire of molecular components essential for an efficient disease resistance response.
Sigma-Aldrich
Uridine, Vetec, reagent grade, 99%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of Ph. Eur., BP, 35 wt. %, contains 10% methanol as stabilizer
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
5-Methyluridine, 97%
Sigma-Aldrich
7-Methylguanosine, ≥90%
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, Vetec, reagent grade, 98%