  • Search Results
  • Cytotoxicity and genotoxicity of zingiberene on different neuron cell lines in vitro.
Cytotoxicity and genotoxicity of zingiberene on different neuron cell lines in vitro.
Cytotechnology (2014-05-08)
Basak Togar, Hasan Turkez, Abdulgani Tatar, Ahmet Hacimuftuoglu, Fatime Geyikoglu
ABSTRACT
The main objective of this study is to investigate the cytotoxic, genotoxic and antioxidant properties of zingiberene (ZBN) in an in vitro rat brain cell culture study. The cytotoxic effect was determined against the rat neuron and N2a neuroblastoma (N2a-NB) cell lines using the 3,(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay, while the antioxidant activity was assessed using the total antioxidant capacity (TAC) and total oxidative stress (TOS) assays. The effects on DNA damage were also evaluated in this study by the single cell gel electrophoresis assay. The results indicated that ZBN has an anti-proliferative activity suppressing the proliferation of N2a-NB cells at concentrations over 50 mg L(-1) and neuron cells at concentrations over 150 mg L(-1). In addition, ZBN treatments at higher doses (≤50 mg L(-1)) led to increases of TOS levels in N2a-NB cell cultures. However 25 mg L(-1) of ZBN treatment caused increases of TAC levels in cultured neuron and N2a-NB cell cultures while ZBN at doses of 10-400 mg L(-1) did not increase the number of total damage score in both cell lines. This study clearly indicates that ZBN has a significant potential to be used as a natural anticancer agent in cultured N2a-NBs.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)