  • Search Results
  • Prevalence and risk factors associated with Clostridium difficile shedding in veal calves in Italy.
Prevalence and risk factors associated with Clostridium difficile shedding in veal calves in Italy.
Anaerobe (2015-02-02)
Chiara Francesca Magistrali, Carmen Maresca, Lucilla Cucco, Luca Bano, Ilenia Drigo, Giovanni Filippini, Annalisa Dettori, Sayra Broccatelli, Giovanni Pezzotti
ABSTRACT
The aim of this study is to describe the prevalence and risk factors of Clostridium difficile shedding in six farms belonging to two companies in Northern Italy. Four hundred and twenty veal calves, randomly selected and individually identified, were sampled three times: at 0-16, 90-120, and 150 days after introduction. C. difficile was isolated at least once from 87 out of the 420 calves (20.7%). The prevalence of shedding was 20.24% at the first sampling and dropped to 0.72% at the second sampling. None of the samples obtained at 150 days tested positive. Sampling of cecal contents and carcass swabs at slaughter was stratified according to the herd of origin of the animals. C. difficile was never isolated at slaughter, excluding a prevalence higher than 3.5% on the basis of previous investigations. Therefore, in this work, the veal calf could not be confirmed as a potential source of C. difficile for the consumer. Eight different ribotypes (RT) have been described, but the vast majority of the isolates (87.8%) belonged to three ribotypes only: RT-078, RT-012 and RT-126, which are also among the most common of the ribotypes detected in humans in Europe. Most isolates, and all the RT-078 isolates, harbored genes coding for toxins A and B, the binary toxin, and showed a deletion in the gene encoding toxin C, suggesting that the veal calf was a reservoir for epidemic hyper-virulent strains. A correlation between age and shedding was found: the odds ratio (OR) ranged from 2.79 for 36-45 days of age to 4.57 for 13-28 days of age. The presence of diarrhea at first sampling was significantly associated with the recovery of C. difficile in feces (OR 3.26). A correlation was found between the administration of antimicrobials and shedding: an increased risk was shown when the number of antimicrobials used was higher than 4 (OR 4.02) or 5-6 (OR 5.83) or when polymyxin E or beta-lactams were administered.
Sigma-Aldrich
D-(−)-Fructose, BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99%
Sigma-Aldrich
D-(−)-Fructose, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, 99.998% trace metals basis