  • Search Results
  • Antidiabetic effects of flavonoids from Sophora flavescens EtOAc extract in type 2 diabetic KK-ay mice.
Antidiabetic effects of flavonoids from Sophora flavescens EtOAc extract in type 2 diabetic KK-ay mice.
Journal of ethnopharmacology (2015-06-09)
Xinzhou Yang, Jing Yang, Chan Xu, Mi Huang, Qi Zhou, Jingnan Lv, Xinhua Ma, Changqiang Ke, Yang Ye, Guangwen Shu, Ping Zhao
ABSTRACT
Bitter and cold Chinese medicines have been long used for the treatment for diabetes mellitus (DM) for thousands of years in China. The roots of Sophora flavescens Ait., one of bitter and cold Chinese medicines commonly used to remove lung heat have been used to counteract DM and exerted good clinical effects for diabetic patients in some folk hospitals in Fujian province, PR China. However, the corresponding active principles and antidiabetic mechanism of this Traditional Chinese Medicine remain unclear. Therefore, in this study, we aim at chemical profiling of the active principles, validating the potential antidiabetic effects of the active EtOAc extract (SF-EtOAc) in vitro and in vivo, and elucidating its probable antidiabetic mechanism as well as evaluating its acute oral toxicity. An off-line semi-preparative LC-NMR and LC-UV-ESI MS protocol was developed to determine the chemical principles of the active EtOAc extract rapidly and unambiguously. The effect of SF-EtOAc on the glucose transporter type 4 (GLUT4) translocation in L6 myotubes was examined. T2DM KK-Ay mice were induced by high-fat diet. SF-EtOAc was orally administration at the dose of 30, 60 and 120 mg/kg/d, for 21 days. Metformin was used as positive control. Body weight, plasma glucose, oral glucose tolerance test, serum insulin and blood-lipid indexes were measured. Phosphorylation of the AMP-activated protein kinase (AMPK) expression in liver was measured. We found that SF-EtOAc significantly improved oral glucose tolerance, increased serum high density lipoprotein cholesterol (HDL-C) and reduced body weight, blood glucose levels and other related blood-lipid indexes. Mechanistically, SF-EtOAc elevated phosphorylation of AMP-activated protein kinase (AMPK) and stimulated membrane translocation of GLUT4. Moreover, it was unveiled that oral median lethal dose (LD50) of SF-EtOAc was more than 7500 mg/kg, suggesting that SF-EtOAc was practically non-toxic for mice. SF-EtOAc improves glucose tolerance, reduces hyperglycemia and resumes insulin levels, at least in part, by activating GLUT4 translocation which may be modulated by AMPK pathway. According to the results of the present study, SF-EtOAc possesses a potent antidiabetic activity and could be used as a safe remedy for the treatment of diabetes.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis