  • Search Results
  • Binding of bis-(2-ethylhexyl) phthalate at the surface of hydrozincite nanocrystals: An example of organic molecules absorption onto nanocrystalline minerals.
Binding of bis-(2-ethylhexyl) phthalate at the surface of hydrozincite nanocrystals: An example of organic molecules absorption onto nanocrystalline minerals.
Journal of colloid and interface science (2015-07-22)
Roberta Sanna, Daniela Medas, Francesca Podda, Carlo Meneghini, Mariano Casu, Pierfranco Lattanzi, Mariano Andrea Scorciapino, Costantino Floris, Carla Cannas, Giovanni De Giudici
ABSTRACT
As a contribution to understand the interactions between mineral surfaces and organic molecules, this study reports an accurate characterization of the bis-(2-ethylhexyl) phthalate (DEHP)-Hydrozincite (DEHP-HY), that has been conduced combining the following techniques: FTIR, NMR, XAS spectroscopies and XRD. XRD patterns indicate that the HY is made of nanocrystals whose size is not influenced by the presence of DEHP. The (1)H NMR analysis of DEHP-HY samples points out the presence of interactions of DEHP with HY. CPMAS NMR analysis suggests that the interaction is operated by ester carbonyl groups while the aliphatic chain, as expected, is not involved. MAS and CPMAS NMR measurements, performed on (13)C ester carbonyl enriched DEHP, allow to demonstrate that there are two ester carbonyl linkage sites interacting at the HY surface: an acid site with a strong link and a second one with weak chemical interactions. Zn K-edge XAS spectroscopy demonstrates that the local atomic structure around Zn in DEHP-HY sample remains essentially unchanged with respect to that of HY. Such a weak structural effect suggests that HY interaction with DEHP is limited to the nanoparticle surface.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Rhein
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium bicarbonate, suitable for cell culture
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bicarbonate, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium sulfate, Vetec, reagent grade, 99%
Sigma-Aldrich
Phthalic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Benzene, ReagentPlus®, thiophene free, ≥99%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Benzene, anhydrous, 99.8%