  • Search Results
  • PLGA-Based Nanoparticles: a Safe and Suitable Delivery Platform for Osteoarticular Pathologies.
PLGA-Based Nanoparticles: a Safe and Suitable Delivery Platform for Osteoarticular Pathologies.
Pharmaceutical research (2015-07-03)
Mathieu Riffault, Jean-Luc Six, Patrick Netter, Pierre Gillet, Laurent Grossin
ABSTRACT
Despite the promising applications of PLGA based particles, studies examining the fate and consequences of these particles after intra-articular administration in the joint are scanty. This study was carried out to evaluate the neutrality of the unloaded delivery system on different articular cell types. To facilitate tracking, we have thus developed a fluorescent core of particles, combined to a hyaluronate shell for cell recognition. Fluorescence pictures were taken at time intervals to assess the internalization and the corresponding inflammatory response was monitored by RT-qPCR and biochemical measurements. After NPs pre-treatment, mesenchymal stem cells (MSCs) were cultured into chondrogenic, adipogenic or osteogenic differentiation media, to investigate if NPs exposure interferes with differentiation ability. Finally, intra-articular injections were performed in healthy rat knees and joint's structure analysed by histological studies. Particles were detected in cytoplasm 8 h after exposure. Internalization led to a slight and reversible increase of inflammatory markers, but lower than in inflammatory conditions. We have confirmed particles exposure minimal neutrality on MSCs pluripotency. Histological exams of joint after intra-articular injections do not demonstrate any side effects of NPs. Our findings suggest that such a delivery platform is well tolerated locally and could be used to deliver active molecules to the joint.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Lactic acid, 85%, FCC