  • Search Results
  • Comparative proteomic analysis of hemolymph proteins from Autographa californica multiple nucleopolyhedrovirus (AcMNPV)-sensitive or -resistant silkworm strains during infections.
Comparative proteomic analysis of hemolymph proteins from Autographa californica multiple nucleopolyhedrovirus (AcMNPV)-sensitive or -resistant silkworm strains during infections.
Comparative biochemistry and physiology. Part D, Genomics & proteomics (2015-08-16)
Jian Xu, Pingbo Zhang, Takahiro Kusakabe, Hiroaki Mon, Zhiqing Li, Li Zhu, Kazuhiro Iiyama, Yutaka Banno, Daisuke Morokuma, Jae Man Lee
ABSTRACT
We reported previously that baculovirus AcMNPV host-ranges in silkworm strains are controlled by a novel third chromosomal locus. To further isolate the potential host factor and uncover the functional pathway involved, in this study we analyzed hemolymph proteins from AcMNPV-resistant or -sensitive silkworm strains infected with baculoviruses. All the protein spots from 2D electrophoresis were characterized by MALDI-TOF MS and further systematically assessed for differentially regulated proteins at different stages of infection. Subsequently, six candidates were selected for functional analysis using Bm5 cells, where the candidates were knocked-down or overexpressed. We observed that mRNA expression levels of beta-N-acetylglucosaminidase and prophenoloxidase subunit 2 are significantly upregulated during AcMNPV infections in Bm5 cells. Ultimately, we found that RNA interference of ribosomal protein RpL34 causes serious damages to cell viability as well as abortive infection, indicating that ribosomal components are essential for productive baculovirus infection.
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C