Merck
Mechanism of scrapie prion precipitation with phosphotungstate anions.
ACS chemical biology (2015-02-20)
Dana J Levine, Jan Stöhr, Lillian E Falese, Julian Ollesch, Holger Wille, Stanley B Prusiner, Jeffrey R Long
ABSTRACT
The phosphotungstate anion (PTA) is widely used to facilitate the precipitation of disease-causing prion protein (PrP(Sc)) from infected tissue for applications in structural studies and diagnostic approaches. However, the mechanism of this precipitation is not understood. In order to elucidate the nature of the PTA interaction with PrP(Sc) under physiological conditions, solutions of PTA were characterized by NMR spectroscopy at varying pH. At neutral pH, the parent [PW12O40](3-) ion decomposes to give a lacunary [PW11O39](7-) (PW11) complex and a single orthotungstate anion [WO4](2-) (WO4). To measure the efficacy of each component of PTA, increasing concentrations of PW11, WO4, and mixtures thereof were used to precipitate PrP(Sc) from brain homogenates of scrapie prion-infected mice. The amount of PrP(Sc) isolated, quantified by ELISA and immunoblotting, revealed that both PW11 and WO4 contribute to PrP(Sc) precipitation. Incubation with sarkosyl, PTA, or individual components of PTA resulted in separation of higher-density PrP aggregates from the neuronal lipid monosialotetrahexosylganglioside (GM1), as observed by sucrose gradient centrifugation. These experiments revealed that yield and purity of PrP(Sc) were greater with polyoxometalates (POMs), which substantially supported the separation of lipids from PrP(Sc) in the samples. Interaction of POMs and sarkosyl with brain homogenates promoted the formation of fibrillar PrP(Sc) aggregates prior to centrifugation, likely through the separation of lipids like GM1 from PrP(Sc). We propose that this separation of lipids from PrP is a major factor governing the facile precipitation of PrP(Sc) by PTA from tissue and might be optimized further for the detection of prions.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)