  • Search Results
  • Understanding the UCST-type transition of P(AAm-co-AN) in H2O and D2O: dramatic effects of solvent isotopes.
Understanding the UCST-type transition of P(AAm-co-AN) in H2O and D2O: dramatic effects of solvent isotopes.
Soft matter (2015-08-08)
Lei Hou, Peiyi Wu
ABSTRACT
The UCST-type transition of poly(acrylamide-co-acrylonitrile) (P(AAm-co-AN)) (molar fraction of AN: 13.3%; PDI = 3.2) in H2O and D2O is explored and compared by applying turbidity, DLS as well as FTIR measurements. The transition temperature of P(AAm-co-AN) in D2O is observed to be almost 10 °C higher than that in H2O at the same concentration, demonstrating a dramatic solvent isotope effect. Such a phenomenon could be rooted from a stronger interaction among polymer chains in D2O than in H2O, as indicated from DLS results. It is also observed in second-derivative analysis of FTIR spectra in the ν(C=O) region, where all C=O groups participate in the formation of inter-/intra-chain hydrogen bonds (C=O···H-N) in D2O while there is still part of relatively "free" C=O groups in H2O. Moreover, we find in the temperature-dependent FTIR spectra that C≡N groups exhibit hydrating behavior while C=O groups present increased inter-/intra-molecular hydrogen bonding interaction (C=O···H-N) upon cooling, revealing the later process to be the driving force of the UCST-type transition.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D