  • Search Results
  • Montmorillonite/graphene oxide/chitosan composite: Synthesis, characterization and properties.
Montmorillonite/graphene oxide/chitosan composite: Synthesis, characterization and properties.
International journal of biological macromolecules (2015-06-07)
Mithilesh Yadav, Sharif Ahmad
ABSTRACT
The present work reports the successful preparation, thermal and mechanical characterization of high performance films of Na(+) montmorillonite (MMT)/graphene oxide (GO)/chitosan (CS) composite using simple solution mixing evaporation method. The formations of films were verified by Fourier transform infrared (FT-IR) spectroscopy, scanning electron microscopy (SEM), X-ray diffraction (XRD) and Raman spectroscopy. The thermal stability and mechanical properties of these films were investigated by thermogravimetric analysis (TGA) and mechanical testing (Instron 8871). The results obtained from these studies revealed that the composites of chitosan, MMT, and graphene oxide were homogeneous in nature. A synergistic effect of MMT and GO reinforcing on chitosan matrix was observed for the first time, in case of 5 wt.% MMT and 1 wt.% GO. The tensile strength of (5 wt.%) MMT/(1 wt.%) GO/CS composite was formed 9±0.23% and 27±0.25% higher than that of the (1 wt.%) GO/CS composite and chitosan, respectively.
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
(Methylcyclopentadienyl)manganese(I) tricarbonyl
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Potassium permanganate, meets USP testing specifications