  • Search Results
  • Effect of Progesterone, Its Hydroxylated and Methylated Derivatives, and Dydrogesterone on Lipid Bilayer Membranes.
Effect of Progesterone, Its Hydroxylated and Methylated Derivatives, and Dydrogesterone on Lipid Bilayer Membranes.
The Journal of membrane biology (2015-04-26)
Rola Abboud, Hélène Greige-Gerges, Catherine Charcosset
ABSTRACT
The interaction of progesterone (PG), 17-hydroxyprogesterone (17-OHPG), 21-hydroxyprogesterone (21-OHPG), medroxyprogesterone (MP), medroxyprogesterone acetate (MPA), and dydrogesterone (DYG), with zwitterionic dipalmitoyl phosphatidylcholine (DPPC) multilamellar liposome, was investigated as a function of drug concentration using Fourier transform infrared spectroscopy and differential scanning calorimetry. The results reveal that progesterone and its derivatives changed the physical properties of the DPPC bilayers by decreasing the main phase-transition temperature (T m) and enthalpy (ΔH m), abolishing the pre-transition and disordering the membrane. From the thermodynamic parameters analysis, we concluded that PG, 21-OHPG, and MPA are localized inside the membrane. Whereas, the insertion of 17-OHPG in the lipid bilayers cannot be excluded in view of the significant decrease in the transition enthalpy at two molar ratios. MP and DYG are rather localized near the polar heads of phospholipids at the interface water-lipid bilayer. PG derivatives increase the membrane fluidity in the order: PG ≈ 21-OHPG ≈ MPA > 17-OHPG > MP ≈ DYG. The distinct effects produced by steroids are discussed in terms of hydrophobicity and chemical structure.
Sigma-Aldrich
1,3-Propanediol, 98%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC