  • Search Results
  • Biosynthesis of membrane dependent proteins in insect cell lysates: identification of limiting parameters for folding and processing.
Biosynthesis of membrane dependent proteins in insect cell lysates: identification of limiting parameters for folding and processing.
Biological chemistry (2015-05-23)
Helmut Merk, Ralf-Bernhardt Rues, Christine Gless, Kerstin Beyer, Fang Dong, Volker Dötsch, Michael Gerrits, Frank Bernhard
ABSTRACT
G protein-coupled receptors, like many other membrane proteins, are notoriously difficult to synthesize in conventional cellular systems. Although expression in insect cells is considered the preferred technique for structural characterizations in particular, inefficient membrane translocation, instability, toxic effects and low yields still pose clear limitations for their production in living cells. Recent studies started to explore alternative strategies for the in vitro production of problematic membrane proteins in cell-free lysates in combination with supplied membranes. We provide a detailed study on the production efficiencies and quality of G protein-coupled receptors, Fab fragments and other proteins synthesized in insect cell lysates containing endogenous microsomes. Effects of different reaction kinetics, redox conditions and sample preparations on the specific activities of synthesized proteins have been analyzed. The extent of glycosylation, membrane translocation and percentages of ligand binding active fractions of synthesized protein samples have been determined. We provide strong evidence that membrane insertion of integral membrane proteins can represent a prime limiting factor for their preparative scale in vitro production. Improved expression protocols resulting into higher production rates yielded more active protein in case of Fab fragments, but not in case of the human endothelin B receptor.
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, Vetec, reagent grade, 99%
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O