  • Search Results
  • Influence of Ligand Shell Composition upon Interparticle Interactions in Multifunctional Nanoparticles.
Influence of Ligand Shell Composition upon Interparticle Interactions in Multifunctional Nanoparticles.
Langmuir : the ACS journal of surfaces and colloids (2015-10-27)
Zachary C Kennedy, Carmen E Lisowski, Dumitru S Mitaru-Berceanu, James E Hutchison
ABSTRACT
The interactions of nanoparticles with biomolecules, surfaces, or other nanostructures are dictated by the nanoparticle's surface chemistry. Thus, far, shortcomings of syntheses of nanoparticles with defined ligand shell architectures have limited our ability to understand how changes in their surface composition influence reactivity and assembly. We report new synthetic approaches to systematically control the number (polyvalency), length, and steric interactions of omega-functionalized (targeting) ligands within an otherwise passivating (diluent) ligand shell. A mesofluidic reactor was used to prepare nanoparticles with the same core diameter for each of the designed ligand architectures. When the targeting ligands are malonamide groups, the nanoparticles assemble via cross-linking in the presence of trivalent lanthanides. We examined the influence of ligand composition on assembly by monitoring the differences in optical properties of the cross-linked and free nanoparticles. Infrared spectroscopy, electron microscopy, and solution small-angle X-ray scattering provided additional insight into the assembly behavior. Lower (less than 33%) malonamide ligand densities (where the binding group extends beyond the periphery of diluent ethylene glycol ligands) produce the strongest optical responses and largest assemblies. Surprisingly, nanoparticles containing a higher surface number of targeting ligand did not produce an optical response or assemble, underscoring the importance of an informed mixed ligand strategy for highest nanoparticle performance.
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
p-Toluenesulfonyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Malonamide, 97%
Sigma-Aldrich
p-Toluenesulfonyl chloride, reagent grade, ≥98%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, Vetec, reagent grade
Sigma-Aldrich
Sodium iodide, puriss. p.a., ≥99.0% (AT)
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3,4-Dihydro-2H-pyran, 97%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes