  • Search Results
  • Optimizing the design and in vitro evaluation of bioreactive glucose oxidase-microspheres for enhanced cytotoxicity against multidrug resistant breast cancer cells.
Optimizing the design and in vitro evaluation of bioreactive glucose oxidase-microspheres for enhanced cytotoxicity against multidrug resistant breast cancer cells.
Colloids and surfaces. B, Biointerfaces (2015-04-22)
Ji Cheng, Qun Liu, Adam J Shuhendler, Andrew M Rauth, Xiao Yu Wu
ABSTRACT
Glucose oxidase (GOX) encapsulated in alginate-chitosan microspheres (GOX-MS) was shown in our previous work to produce reactive oxygen species (ROS) in situ and exhibit anticancer effects in vitro and in vivo. The purpose of present work was to optimize the design and thus enhance the efficacy of GOX-MS against multidrug resistant (MDR) cancer cells. GOX-MS with different mean diameters of 4, 20 or 140 μm were prepared using an emulsification-internal gelation-adsorption-chitosan coating method with varying compositions and conditions. The GOX loading efficiency, loading level, relative bioactivity of GOX-MS, and GOX leakage were determined and optimal chitosan concentrations in the coating solution were identified. The influence of particle size on cellular uptake, ROS generation, cytotoxicity and their underlying mechanisms was investigated. At the same GOX dose and incubation time, smaller sized GOX-MS produced larger amounts of H2O2 in cell culture medium and greater cytotoxicity toward murine breast cancer MDR (EMT6/AR1.0) and wild type (EMT6/WT) cells. Fluorescence and confocal laser scanning microscopy revealed significant uptake of small sized (4 μm) GOX-MS by both MDR and WT cells, but no cellular uptake of large (140 μm) GOX-MS. The GOX-MS were equally effective in killing both MDR cells and WT cells. The cytotoxicity of the GOX formulations was positively correlated with membrane damage and lipid peroxidation. GOX-MS induced greater membrane damage and lipid peroxidation in MDR cells than the WT cells. These results suggest that the optimized, small micron-sized GOX-MS are highly effective against MDR breast cancer cells.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, 97%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, Vetec, reagent grade, 99%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis