  • Search Results
  • Regulation of the Melanocortin-Sensitive Adenylate Cyclase System by N-Acylated Peptide 71-82 of Type 4 Melanocortin Receptor.
Regulation of the Melanocortin-Sensitive Adenylate Cyclase System by N-Acylated Peptide 71-82 of Type 4 Melanocortin Receptor.
Bulletin of experimental biology and medicine (2015-11-26)
A O Shpakov, K V Derkach, E A Shpakova
ABSTRACT
The peptides structurally corresponding in to cytoplasmic loops of G protein-coupled receptors (GPCR) are able to control functional activity of homologous receptors and the corresponding signaling pathways. Modification of these peptides with hydrophobic radicals enhances their biological activity due to penetration of lipophilic derivatives through the membrane and anchoring near their targets, GPCR. We synthesized an N-palmitoylated peptide Palm-Val-[Lys-Asn-Lys-Asn-Leu-His-Ser-Pro-(Nle)-Tyr-Phe-Phe71-82]-amide-Palm-Val-(71-82) structurally corresponding to cytoplasmic loop 1 of melanocortin 4 receptor (M4R). We found that in micromolar concentrations it very effectively suppresses stimulation of basal adenylate cyclase activity and basal level of GppNHp binding of heterotrimeric G proteins produced by THIQ and α-melanocyte stimulating hormone (α-MSH), agonists of M4R homologous to the peptide, in synaptosomal membranes of rat brain. The peptide Palm-Val-(71-82) also reduced, albeit to a significantly less extent, stimulation of adenylate cyclase and G-proteins by M3R agonist of γ-MSH, due to high homology of the peptide primary structure to M3R cytoplasmic loop 1. The synthesized peptide with activity of M4R/M3R antagonist can be used for the development of regulators of M4R and M3R and the corresponding biochemical and physiological processes.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
1,2,3,4-Tetrahydroisoquinoline, 95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)