  • Search Results
  • Augmented pentose phosphate pathway plays critical roles in colorectal carcinomas.
Augmented pentose phosphate pathway plays critical roles in colorectal carcinomas.
Oncology (2015-01-17)
Norisuke Shibuya, Ken-ichi Inoue, Genki Tanaka, Kazumi Akimoto, Keiichi Kubota
ABSTRACT
Glycolysis and the pentose phosphate pathway (PPP) are preferentially activated in cancer cells. Accumulating evidence indicated the significance of the altered glucose metabolism in cancer, but the implication for oncotherapy remains unclear. Here we report that the synthesis of glycolytic and PPP enzymes is almost ubiquitously augmented in colorectal carcinoma (CRC) specimens. The mammalian target of rapamycin (mTOR) inhibitor INK128 (300 nM) and phytochemical Avemar (1 mg/ml) inhibited the synthesis of PPP enzymes in CRC cell lines. INK128 (150-600 nM) and resveratrol (75-300 μM) inhibited aerobic glycolysis in the cell lines. INK128 (300 nM) and Avemar (1 mg/ml) decreased the NADPH/NADP(+) ratio as well as the GSH/GSSG ratio in the cell lines. Finally, per os administration of INK128 (0.8 mg/kg) or Avemar (1 g/kg) suppressed tumor growth and delayed tumor formation by transplantable CRC specimens derived from patients. Taken together, pharmacological inhibition of the mTOR-PPP axis is a promising therapeutic strategy against CRCs.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Spermidine, BioReagent, for molecular biology, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Sucrose, ACS reagent