  • Search Results
  • Improving Antitumor Activity with N-Trimethyl Chitosan Entrapping Camptothecin in Colon Cancer and Lung Cancer.
Improving Antitumor Activity with N-Trimethyl Chitosan Entrapping Camptothecin in Colon Cancer and Lung Cancer.
Journal of nanoscience and nanotechnology (2015-12-31)
Lu Sun, Xingyi Li, Zhengguang Li, Zhiyong Li, Maling Gou, Zhiyong Qian, Feng Peng
ABSTRACT
Camptothecin (CPT) exerts very strong antitumor activities by suppressing the activity of DNA topoisomerase I, but its application is greatly limited owing to its low solubility and the instability of the active lactone form. To overcome this bottleneck, we prepared the novel camptothecin nanocolloids based on N-trimethyl chitosan (CPT-TMC) to efficiently administer CPT systemically. In this study, we investigated the antitumor activity of CPT-TMC against both colon cancer and lung cancer. In vitro cell experiments both CPT and CPT-TMC significantly inhibited the growth of CT26 cells and LL/2 cells, but no statistical difference was observed between CPT-TMC and CPT. In vivo studies, CPT-TMC more effectively inhibited tumor growth and prolonged survival time than CPT both in the CT26 colon carcinoma subcutaneous model and in the LL/2 Lewis lung carcinoma subcutaneous model. In addition, results of PCNA and CD31 immunohistochemical staining of tumor tissues also confirmed the improved antitumor effect of CPT-TMC. These findings suggest that N-trimethyl chitosan could increase the antitumor effect of CPT. Consequently, CPT delivery by N-trimethyl chitosan is a potential approach for effective treatment of cancer.
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder