  • Search Results
  • Antibody structural integrity of site-specific antibody-drug conjugates investigated by hydrogen/deuterium exchange mass spectrometry.
Antibody structural integrity of site-specific antibody-drug conjugates investigated by hydrogen/deuterium exchange mass spectrometry.
Analytical chemistry (2015-05-06)
Lucy Yan Pan, Oscar Salas-Solano, John F Valliere-Douglass
ABSTRACT
We present the results of a hydrogen/deuterium exchange mass spectrometric (HDX-MS) investigation of an antibody-drug conjugate (ADC) comprised of drug-linkers conjugated to cysteine residues that have been engineered into heavy chain (HC) fragment crystallizable (Fc) domain at position 239. A side-by-side comparison of the HC Ser239 wild type (wt) monoclonal antibody (mAb) and the engineered Cys239 mAb indicates that site directed mutagenesis of Ser239 to cysteine has no impact on the HDX kinetics of the mAb. According to the crystal structure of a homologous immunoglobulin G1 (IgG1) antibody (PDB: 1HZH ), the backbone amide of Ser239 is hydrogen-bonded to Val264 backbone amide in the wt-mAb studied here. Replacing Ser239 with a Cys residue does not alter the exchange kinetics of the backbone amide of Val264 suggesting that either Ser or Cys at position 239 has similar amide-hydrogen bonding with Val264. However, a small segment in CH2 domain of the ADC ((264)VDVS) was found to have a slightly increased HDX rate compared to the wt- and C239-mAb constructs. The slightly increased HDX rate of the segment (264)VDVS in ADCs indicates that the further modification of Cys239 with drug-linkers only attenuates the local backbone amide hydrogen-bonding network between Cys239 and Val264. All other regions which are proximal to the site of drug conjugation are unaffected. The results demonstrate that the site-specific drug conjugation at the engineered Cys residue at the position 239 of HC does not impact the structural integrity of antibodies. The results also highlight the utility of applying HDX-MS to ADCs to gain a molecular level insight into the impact of site-specific conjugation technologies on the higher-order structure (HOS) of mAbs. The methodology can be applied generally to site-specific ADC modalities to understand the individual contributions of site-mutagenesis and drug-linker conjugation on the HOS of therapeutic candidate ADCs.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
2-(4-Biphenylyl)-5-phenyl-1,3,4-oxadiazole, 98%
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Vetec, reagent grade, 99%