  • Search Results
  • Glycan complexity dictates microbial resource allocation in the large intestine.
Glycan complexity dictates microbial resource allocation in the large intestine.
Nature communications (2015-06-27)
Artur Rogowski, Jonathon A Briggs, Jennifer C Mortimer, Theodora Tryfona, Nicolas Terrapon, Elisabeth C Lowe, Arnaud Baslé, Carl Morland, Alison M Day, Hongjun Zheng, Theresa E Rogers, Paul Thompson, Alastair R Hawkins, Madhav P Yadav, Bernard Henrissat, Eric C Martens, Paul Dupree, Harry J Gilbert, David N Bolam
ABSTRACT
The structure of the human gut microbiota is controlled primarily through the degradation of complex dietary carbohydrates, but the extent to which carbohydrate breakdown products are shared between members of the microbiota is unclear. We show here, using xylan as a model, that sharing the breakdown products of complex carbohydrates by key members of the microbiota, such as Bacteroides ovatus, is dependent on the complexity of the target glycan. Characterization of the extensive xylan degrading apparatus expressed by B. ovatus reveals that the breakdown of the polysaccharide by the human gut microbiota is significantly more complex than previous models suggested, which were based on the deconstruction of xylans containing limited monosaccharide side chains. Our report presents a highly complex and dynamic xylan degrading apparatus that is fine-tuned to recognize the different forms of the polysaccharide presented to the human gut microbiota.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
L-(+)-Arabinose, ≥99%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
L-(−)-Fucose, ≥99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Hematin porcine
Sigma-Aldrich
L-(+)-Arabinose, 99%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99%
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture