  • Search Results
  • Proliposomes containing a bile salt for oral delivery of Ginkgo biloba extract: Formulation optimization, characterization, oral bioavailability and tissue distribution in rats.
Proliposomes containing a bile salt for oral delivery of Ginkgo biloba extract: Formulation optimization, characterization, oral bioavailability and tissue distribution in rats.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-06-28)
Bin Zheng, Lirong Teng, Gaoyang Xing, Ye Bi, Shuang Yang, Fei Hao, Guodong Yan, Xinmei Wang, Robert J Lee, Lesheng Teng, Jing Xie
ABSTRACT
Proliposomes containing a bile salt were developed to improve the oral bioavailability of Ginkgo biloba extract (GbE). GbE loaded proliposomes (P-GbE) were successfully prepared by spray drying method. The formulation was optimized using the response surface methodology. FE-SEM, DSC, and FT-IR were used to study the surface morphology and molecular state of proliposomes, and demonstrated key interactions between the formulation ingredients. In vitro studies showed delayed release and enhanced dissolution of Ginkgo flavonoids and terpene lactones from GbE proliposomes. Proliposomes significantly enhanced GbE absorption in the gastrointestinal tract and decreased its elimination. The bioavailabilities of quercetin, kaempferol, isorhmnetin, ginkgolide A, ginkgolide B and ginkgolide C from proliposomes relative to the control were 245%, 211%, 264%, 203%, 333%, and 294%, respectively. Proliposomes were shown to selectively deliver GbE to critical target tissues. In conclusion, development of proliposomes formulation for GbE solved the problem of its poor oral bioavailability, prolonged its duration of action, and increased drug distribution in critical tissues, especially in the brain, therefore, warrant further investigation.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
SAFC
Sodium deoxycholate
Sigma-Aldrich
Isorhamnetin, ≥95.0% (HPLC)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG