  • Search Results
  • Availability of extracellular matrix biopolymers and differentiation state of human mesenchymal stem cells determine tissue-like growth in vitro.
Availability of extracellular matrix biopolymers and differentiation state of human mesenchymal stem cells determine tissue-like growth in vitro.
Biomaterials (2015-05-20)
Manuela Herklotz, Marina C Prewitz, Cécile M Bidan, John W C Dunlop, Peter Fratzl, Carsten Werner
ABSTRACT
To explore the space-filling growth of adherent mesenchymal stem cells (MSC) into tissue-like structures in vitro, human bone marrow derived MSC were exposed to fibronectin-coated, millimeter-sized, triangular channels casted in poly(dimethyl siloxane) carriers. The results revealed that the three dimensional (3D) growth of MSC differs in dependence on differentiation status and availability of extracellular matrix (ECM) proteins: Massive 3D structure formation was observed for MSC under pro-osteogenic stimulation but not for undifferentiated MSC nor for MSC under pro-adipogenic stimulation; boosting cellular matrix secretion and addition of soluble ECM proteins caused extensive 3D tissue formation of undifferentiated MSC. The reported findings may contribute to bridge the gap between in vitro and in vivo analyses and guide the application of MSC in tissue replacement approaches.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Picric acid solution, 0.9-1.1% (alkalimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, Vetec, reagent grade, 99%
Sigma-Aldrich
DAPI, for nucleic acid staining