  • Search Results
  • Redox properties of the nitronyl nitroxide antioxidants studied via their reactions with nitroxyl and ferrocyanide.
Redox properties of the nitronyl nitroxide antioxidants studied via their reactions with nitroxyl and ferrocyanide.
Free radical research (2015-03-20)
A A Bobko, V V Khramtsov
ABSTRACT
Nitronyl nitroxides (NNs) are the paramagnetic probes that are capable of scavenging physiologically relevant reactive oxygen (ROS) and nitrogen (RNS) species, namely superoxide, nitric oxide (NO), and nitroxyl (HNO). NNs are increasingly considered as potent antioxidants and potential therapeutic agents. Understanding redox chemistry of the NNs is important for their use as antioxidants and as paramagnetic probes for discriminative detection of NO and HNO by electron paramagnetic resonance (EPR) spectroscopy. Here we investigated the redox properties of the two most commonly used NNs, including determination of the equilibrium and rate constants of their reduction by HNO and ferrocyanide, and reduction potential of the couple NN/hydroxylamine of nitronyl nitroxide (hNN). The rate constants of the reaction of the NNs with HNO were found to be equal to (1-2) × 10(4) M(-1)s(- 1) being close to the rate constants of scavenging superoxide and NO by NNs. The reduction potential of the NNs and iminonitroxides (INs, product of NNs reaction with NO) were calculated based on their reaction constants with ferrocyanide. The obtained values of the reduction potential for NN/hNN (E'0 ≈ 285 mV) and IN/hIN (E' ≈ 495 mV) are close to the corresponding values for vitamin C and vitamin E, correspondingly. The "balanced" scavenging rates of the NNs towards superoxide, NO, and HNO, and their low reduction potential being thermodynamically close to the bottom of the pecking order of oxidizing radicals, might be important factors contributing into their antioxidant activity.
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, 99.998% trace metals basis
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%