  • Search Results
  • Polyphosphates form antigenic complexes with platelet factor 4 (PF4) and enhance PF4-binding to bacteria.
Polyphosphates form antigenic complexes with platelet factor 4 (PF4) and enhance PF4-binding to bacteria.
Thrombosis and haemostasis (2015-08-01)
Sven Brandt, Krystin Krauel, Miriam Jaax, Thomas Renné, Christiane A Helm, Sven Hammerschmidt, Mihaela Delcea, Andreas Greinacher
ABSTRACT
Short chain polyphosphates (polyP) are pro-coagulant and pro-inflammatory platelet released inorganic polymers. The platelet chemokine platelet factor 4 (PF4) binds to lipid A on bacteria, inducing an antibody mediated host defense mechanism, which can be misdirected against PF4/heparin complexes leading to the adverse drug reaction heparin-induced thrombocytopenia (HIT). Here, we demonstrate that PF4 complex formation with soluble short chain polyP contributes to host defense mechanisms. Circular dichroism spectroscopy and isothermal titration calorimetry revealed that PF4 changed its structure upon binding to polyP in a similar way as seen in PF4/heparin complexes. Consequently, PF4/polyP complexes exposed neoepitopes to which human anti-PF4/heparin antibodies bound. PolyP enhanced binding of PF4 to Escherichia coli, hereby facilitating bacterial opsonisation and, in the presence of human anti-PF4/polyanion antibodies, phagocytosis. Our study indicates a role of polyP in enhancing PF4-mediated defense mechanisms of innate immunity.
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%