  • Search Results
  • Aerosol Route to Administer Teicoplanin in Mechanical Ventilation: In Vitro Study, Lung Deposition and Pharmacokinetic Analyses in Pigs.
Aerosol Route to Administer Teicoplanin in Mechanical Ventilation: In Vitro Study, Lung Deposition and Pharmacokinetic Analyses in Pigs.
Journal of aerosol medicine and pulmonary drug delivery (2015-01-24)
Antoine Guillon, Emmanuelle Mercier, Philippe Lanotte, Eve Haguenoer, François Darrouzain, Céline Barc, Pierre Sarradin, Mustapha Si-Tahar, Nathalie Heuzé-Vourc'h, Patrice Diot, Laurent Vecellio
ABSTRACT
Glycopeptides given intravenously achieve low airway concentrations. Nebulization of teicoplanin may be an efficient way of delivering a high concentration of this antibiotic to the lung. This multistep study assessed the feasibility of teicoplanin nebulization during mechanical ventilation by evaluating: the stability of its antibiotic effect; epithelial tolerance; lung deposition and systemic absorption in ventilated pigs. Nebulized and non-nebulized teicoplanin activity was tested on Staphylococcus aureus cultures. The cytotoxic effect of teicoplanin on human respiratory epithelial cells was assessed by measuring lactate dehydrogenase activity released, cell viability, and transepithelial electrical resistance. Volume median diameter of particles of nebulized teicoplanin was measured by laser diffraction during mechanical ventilation. The deposited mass of teicoplanin nebulized with a vibrating mesh nebulizer in ventilated piglets was assessed by scintigraphy. Blood pharmacokinetics of teicoplanin administered either intravenously or by nebulization was compared. No decrease of antibiotic activity was observed after nebulization. In vitro cytotoxicity of teicoplanin was only observed with 1000 times the dose recommended for intravenous administration. Volume median diameter of particles was 2.5±0.1 μm. Of the initial nebulizer charge of teicoplanin, 24±7% was present in the lungs of ventilated pigs after the nebulization. Amount absorbed in blood was low (3.4%±0.9%) after nebulization, and blood stream elimination half-life value was 25.4 h. Teicoplanin was administered efficiently by nebulization during mechanical ventilation, without any effect on its pharmacological properties or any cytotoxicity. The pharmacokinetic parameters are promising in view of its time-dependent killing process. All the results of our multi-step study highlighted the potential of teicoplanin to be nebulized during mechanical ventilation.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
MTT Formazan, powder
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
MTT Formazan, MTT reduction end product
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl