  • Search Results
  • Role of Hydrogen Bonding and Polyanion Composition in the Formation of Lipid Bilayers on Top of Polyelectrolyte Multilayers.
Role of Hydrogen Bonding and Polyanion Composition in the Formation of Lipid Bilayers on Top of Polyelectrolyte Multilayers.
Langmuir : the ACS journal of surfaces and colloids (2015-07-15)
E Diamanti, L Cuellar, D Gregurec, S E Moya, E Donath
ABSTRACT
The self-assembly of mixed vesicles of zwitterionic phosphatidylcholine (PC) and anionic phosphatidylserine (PS) phospholipids on top of polyelectrolyte multilayers (PEMs) of poly(allylamine hydrochloride) (PAH), as a polycation, and polystyrenesulfonate (PSS), as a polyanion, is investigated as a function of the vesicle composition by means of the quartz crystal microbalance with dissipation (QCM-D), cryo-transmission electron microscopy (Cryo-TEM), atomic force microscopy (AFM), and atomic force spectroscopy (AFS). Vesicles with molar percentages of PS between 50% and 70% result in the formation of lipid bilayers on top of the PEMs. Vesicles with over 50% of PC or over 80% of PS do not assembly into bilayers. AFS studies performed with a PAH-modified cantilever approaching and retracting from the lipid assemblies reveal that the main interaction between PAH and the lipids takes place through hydrogen bonding between the amine groups of PAH and the carboxylate and phosphate groups of PS and with the phosphate groups of PC. The interaction of PAH with PS is much stronger than with PC. AFS measurements on assemblies with 50% PC and 50% PS revealed similar adhesion forces to pure PS assemblies, but the PAH chains can reorganize much better on the lipids as a consequence of the presence of PC. QCM-D experiments show that vesicles with a lipid composition of 50% PC and 50% PS do not form bilayers if PSS is replaced by alginate (Alg) or poly(acrylic acid) (PAA).
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N