  • Search Results
  • Epsilon-caprolactone modified polyethylenimine for highly efficient antigen delivery and chemical exchange saturation transfer functional MR imaging.
Epsilon-caprolactone modified polyethylenimine for highly efficient antigen delivery and chemical exchange saturation transfer functional MR imaging.
Biomaterials (2015-05-03)
Yan Tan, Lisi Xie, Zhiyong Wang, Na Zhang, Chao Zou, Zhi-ying Chen, Xin Liu, Jian Lu, Gang Liu, Hairong Zheng
ABSTRACT
Serious challenges remain in immunotherapy, such as to effectively deliver antigens and non-invasively monitor biological processes. Therefore, we aim to design a useful nanocarrier for enhancing antigen-specific immune response and performing functional molecular imaging guided immunotherapy. To achieve this goal, ε-caprolactone modified polyethylenimine (PEI-CL) was prepared based on the ring-opening reaction. This novel macromolecule served as an optimal vehicle that efficiently bound ovalbumin (OVA) antigen forming a controlled nanostructure through electronic self-assembly and resulted in a substantially increased cellular uptake. In vivo, PEI-CL/OVA nanovaccine stimulated inflammatory cytokine production and promoted proliferation of OVA-specific T cells. Moreover, due to the unique chemical and physical properties, PEI-CL was demonstrated as a chemical exchange saturation transfer contrast agent for magnetic resonance imaging. In conclusion, the PEI-CL has synergistic advantages in the effective delivery of antigen and non-invasive imaging for tracking immunotherapeutic cells.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D