  • Search Results
  • Incremental amounts of ground flaxseed decrease milk yield but increase n-3 fatty acids and conjugated linoleic acids in dairy cows fed high-forage diets(1).
Incremental amounts of ground flaxseed decrease milk yield but increase n-3 fatty acids and conjugated linoleic acids in dairy cows fed high-forage diets(1).
Journal of dairy science (2015-05-11)
T L Resende, J Kraft, K J Soder, A B D Pereira, D E Woitschach, R B Reis, A F Brito
ABSTRACT
The objective of this study was to investigate the effect of incremental amounts of ground flaxseed (GFX) on milk yield and concentrations and yields of milk components, milk fatty acids (FA) profile, ruminal metabolism, and nutrient digestibility in dairy cows fed high-forage diets. Twelve multiparous Jersey cows averaging (mean ± SD) 112±68d in milk and 441±21kg of body weight and 8 primiparous Jersey cows averaging 98±43d in milk and 401±43kg of body weight were randomly assigned to treatment sequences in a replicated 4×4 Latin square design. Each period lasted 21d with 14d for diet adaptation and 7d for data and sample collection. Treatments were fed as a total mixed ration (63:37 forage-to-concentrate ratio) with corn meal and soybean meal replaced by incremental levels (i.e., 0, 5, 10, or 15% diet dry matter) of GFX. The ruminal molar proportions of acetate and butyrate decreased linearly with GFX supplementation, whereas the ruminal molar proportion of propionate increased linearly resulting in decreased acetate-to-propionate ratio. Apparent total-tract digestibilities of nutrients either decreased (dry matter) or tended to decrease (organic matter, neutral detergent fiber, acid detergent fiber) linearly in cows fed GFX. Milk yield decreased linearly in cows fed increasing amounts of GFX, which is explained by the linear reduction in dry matter intake. Except for the concentrations of milk protein and urea N, which decreased linearly with GFX supplementation, no other changes in the concentration of milk components were observed. However, yields of milk protein and fat decreased linearly with GFX supplementation. The linear decrease in the yields of milk fat and protein are explained by reduced milk yield, whereas that in milk urea N is explained by decreased crude protein intake. No treatment effects were observed for plasma urea N and nonesterified fatty acids, serum cortisol, and body weight change. Milk odd- and branched-chain FA and saturated FA decreased linearly with GFX supplementation. Milk trans-11 18:1, α-linolenic acid, cis-9,trans-11 18:2, and the sum of n-3 FA all increased linearly and quadratically, whereas the milk ratio of n-6 to n-3 decreased linearly in cows fed GFX. Overall, compared with the control diet (0% GFX), the diet with 15% GFX supplementation resulted in the lowest milk yield but highest milk proportions and yields (data not shown) of cis-9,trans-11 18:2 and n-3 FA.
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, Vetec, reagent grade, 99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%