  • Search Results
  • Development of a Method for the Analysis of Multiclass Antibiotic Residues in Milk Using QuEChERS and Liquid Chromatography-Tandem Mass Spectrometry.
Development of a Method for the Analysis of Multiclass Antibiotic Residues in Milk Using QuEChERS and Liquid Chromatography-Tandem Mass Spectrometry.
Foodborne pathogens and disease (2015-08-11)
Yuan-long Wang, Zhen-min Liu, Jing Ren, Ben-heng Guo
ABSTRACT
A precise and simplified method of sample preparation for the simultaneous quantification of the antibiotics β-lactam, macrolide, tetracycline, sulfonamide, and quinolone in bovine milk was developed. The central composite design of response surface methodology was used to design and optimize the method for the determination of six different antibiotic residues in milk. The recovery of each antibiotic was studied using a quick, easy, cheap, effective, rugged, and safe (QuEChERS) method. Octadecylsilane (C18), primary secondary amine (PSA), and sodium acetate (Na acetate) were the main factors affecting the recovery of each antibiotic. After optimization, the maximum predicted recovery rate was 84.18% for erythromycin under the optimized conditions of 101.20 mg C18, 52.00 mg PSA, and 1.01 g Na acetate. The recovery rates of the five other antibiotic residues ranged from 86.09% to 115.99%. The results suggested that modified QuEChERS could effectively be implemented in the analysis of antibiotic residues in milk.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis