  • Search Results
  • Design of Chitosan-Grafted Carbon Nanotubes: Evaluation of How the -OH Functional Group Affects Cs+ Adsorption.
Design of Chitosan-Grafted Carbon Nanotubes: Evaluation of How the -OH Functional Group Affects Cs+ Adsorption.
Marine drugs (2015-05-27)
Shubin Yang, Dadong Shao, Xiangke Wang, Guangshun Hou, Masaaki Nagatsu, Xiaoli Tan, Xuemei Ren, Jitao Yu
ABSTRACT
In order to explore the effect of -OH functional groups in Cs+ adsorption, we herein used the low temperature plasma-induced grafting method to graft chitosan onto carbon nanotubes (denoted as CTS-g-CNTs), as raw-CNTs have few functional groups and chitosan has a large number of -OH functional groups. The synthesized CTS-g-CNT composites were characterized using different techniques. The effect of -OH functional groups in the Cs+ adsorption process was evaluated by comparison of the adsorption properties of raw-CNTs with and without grafting chitosan. The variation of environmental conditions such as pH and contact time was investigated. A comparison of contaminated seawater and simulated groundwater was also evaluated. The results indicated that: (1) the adsorption of Cs+ ions was strongly dependent on pH and the competitive cations; (2) for CNT-based material, the -OH functional groups have a positive effect on Cs+ removal; (3) simulated contaminated groundwater can be used to model contaminated seawater to evaluate the adsorption property of CNTs-based material. These results showed direct observational evidence on the effect of -OH functional groups for Cs+ adsorption. Our findings are important in providing future directions to design and to choose effective material to remedy the removal of radioactive cesium from contaminated groundwater and seawater, crucial for public health and the human social environment.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥98%
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Cesium chloride, Vetec, reagent grade, ≥99%
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis