Merck
  • Postmortem distribution of MAB-CHMINACA in body fluids and solid tissues of a human cadaver.
Postmortem distribution of MAB-CHMINACA in body fluids and solid tissues of a human cadaver.
Forensic toxicology (2015-08-11)
Koutaro Hasegawa, Amin Wurita, Kayoko Minakata, Kunio Gonmori, Hideki Nozawa, Itaru Yamagishi, Kanako Watanabe, Osamu Suzuki
ABSTRACT
During the latter part of 2014, we experienced an autopsy case in which 5-fluoro-ADB, one of the most dangerous synthetic cannabinoids, was identified and quantitated in solid tissues and in three herbal blend products [Forensic Toxicol (2015) 33:112-121]. At that time, although we suspected that there may be some drug(s) other than 5-fluoro-ADB in the herbal products, all trials to find it/them were unsuccessful. Subsequently, we carefully re-examined the presence of other synthetic cannabinoid(s) in the above herbal blend products using accurate mass spectrometry and found two new compounds, 5-fluoro-ADB-PINACA and MAB-CHMINACA (Forensic Toxicol. doi: 10.1007/s 11419-015-0264-y). In the present communication, we report the distribution of MAB-CHMINACA in body fluids and solid tissue specimens collected from the same deceased individual (kept frozen at -80 °C) as described above for demonstration of 5-fluoro-ADB. Unexpectedly, unchanged MAB-CHMINACA could be identified and quantitated in whole blood and in pericardial fluid specimens, but it was below the detection limit (0.1 ng/ml) in the urine specimen. A higher concentration of MAB-CHMINACA could be found in all of the nine solid tissues; the highest concentration of MAB-CHMINACA was found in the liver (156 ng/g), followed by the kidney, pancreas and so on. The compounds were detected in all nine solid tissues; their levels were generally higher than those in the whole blood and pericardial fluid. Contrary to expectations, the concentration of MAB-CHMINACA in the adipose tissue was relatively low. Our results show that the victim smoked one of the three herbal blend products containing both MAB-CHMINACA and 5-fluoro-ADB, resulting in the coexistence of both compounds. It should be concluded that 5-fluoro-ADB and MAB-CHMINACA synergically exerted their toxicities, leading to death after a short interval. The differences in the distribution of 5-fluoro-ADB and MAB-CHMINACA among the cadaver specimens were also discussed in view of the structures of both compounds. To our knowledge, this is the first report to demonstrate MAB-CHMINACA in biological/human specimens.
Sigma-Aldrich
Octadecylsilane, 97%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, Vetec, reagent grade