  • Search Results
  • Phylogenetic relationships and the evolution of BMP4 in triggerfishes and filefishes (Balistoidea).
Phylogenetic relationships and the evolution of BMP4 in triggerfishes and filefishes (Balistoidea).
Molecular phylogenetics and evolution (2015-09-27)
Charlene L McCord, Mark W Westneat
ABSTRACT
The triggerfishes (family Balistidae) and filefishes (family Monacanthidae) comprise a charismatic superfamily (Balistoidea) within the diverse order Tetraodontiformes. This group of largely marine fishes occupies an impressive ecological range across the world's oceans, and is well known for its locomotor and feeding diversity, unusual body shapes, small genome size, and ecological and economic importance. In order to investigate the evolutionary history of these important fish families, we used multiple phylogenetic methods to analyze molecular data from 86 species spanning the extant biodiversity of Balistidae and Monacanthidae. In addition to three gene regions that have been used extensively in phylogenetic analyses, we include sequence data for two mitochondrial regions, two nuclear markers, and the growth factor gene bmp4, which is involved with cranial development. Phylogenetic analyses strongly support the monophyly of the superfamily Balistoidea, the sister-family relationship of Balistidae and Monacanthidae, as well as three triggerfish and four filefish clades that are well resolved. A new classification for the Balistidae is proposed based on phylogenetic groups. Bayesian topology, as well as the timing of major cladogenesis events, is largely congruent with previous hypotheses of balistid phylogeny. However, we present a novel topology for major clades in the filefish family that illustrate the genera Aluterus and Stephanolepis are more closely related than previously posited. Molecular rates suggest a Miocene and Oligocene origin for the families Balistidae and Monacanthidae, respectively, and significant divergence of species in both families within the past 5 million years. A second key finding of this study is that, relative to the other protein-coding gene regions in our DNA supermatrix, bmp4 shows a rapid accumulation of both synonymous and non-synonymous substitutions, especially within the family Monacanthidae. Overall substitution patterns in bmp4 support the hypothesis of stabilizing selection during the evolutionary history of regulatory genes, with a small number of isolated examples of accelerated non-synonymous changes detected in our phylogeny.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%