  • Search Results
  • Towards clinical application of pronuclear transfer to prevent mitochondrial DNA disease.
Towards clinical application of pronuclear transfer to prevent mitochondrial DNA disease.
Nature (2016-06-10)
Louise A Hyslop, Paul Blakeley, Lyndsey Craven, Jessica Richardson, Norah M E Fogarty, Elpida Fragouli, Mahdi Lamb, Sissy E Wamaitha, Nilendran Prathalingam, Qi Zhang, Hannah O'Keefe, Yuko Takeda, Lucia Arizzi, Samer Alfarawati, Helen A Tuppen, Laura Irving, Dimitrios Kalleas, Meenakshi Choudhary, Dagan Wells, Alison P Murdoch, Douglass M Turnbull, Kathy K Niakan, Mary Herbert
ABSTRACT
Mitochondrial DNA (mtDNA) mutations are maternally inherited and are associated with a broad range of debilitating and fatal diseases. Reproductive technologies designed to uncouple the inheritance of mtDNA from nuclear DNA may enable affected women to have a genetically related child with a greatly reduced risk of mtDNA disease. Here we report the first preclinical studies on pronuclear transplantation (PNT). Surprisingly, techniques used in proof-of-concept studies involving abnormally fertilized human zygotes were not well tolerated by normally fertilized zygotes. We have therefore developed an alternative approach based on transplanting pronuclei shortly after completion of meiosis rather than shortly before the first mitotic division. This promotes efficient development to the blastocyst stage with no detectable effect on aneuploidy or gene expression. After optimization, mtDNA carryover was reduced to <2% in the majority (79%) of PNT blastocysts. The importance of reducing carryover to the lowest possible levels is highlighted by a progressive increase in heteroplasmy in a stem cell line derived from a PNT blastocyst with 4% mtDNA carryover. We conclude that PNT has the potential to reduce the risk of mtDNA disease, but it may not guarantee prevention.
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture